Assay Details
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122093.2 | XP_014977579.2 | 5988 | 69 | ||
XM_015122094.2 | XP_014977580.2 | 5547 | 69 | |||
XM_028838266.1 | XP_028694099.1 | 5722 | 69 | |||
XM_028838267.1 | XP_028694100.1 | 5724 | 69 | |||
XM_028838268.1 | XP_028694101.1 | 5724 | 69 | |||
XM_028838269.1 | XP_028694102.1 | 5724 | 69 | |||
XM_028838270.1 | XP_028694103.1 | 5725 | 69 | |||
XM_028838271.1 | XP_028694104.1 | 5725 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
