Assay Details
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111268.2 | XP_014966754.2 | 1570 | 96 | ||
XM_015111283.2 | XP_014966769.2 | 1345 | 96 | |||
XM_028828334.1 | XP_028684167.1 | 1422 | 96 | |||
XM_028828341.1 | XP_028684174.1 | 1466 | 96 | |||
GenBank mRNA | JU333953.1 | - | 1528 | 96 | ||
JU475136.1 | - | 1515 | 96 | |||
JV464501.1 | - | 1523 | 96 | |||
JV667370.1 | - | 1406 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
