Assay Details
Gene Symbol: VSIG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152928.2 | XP_015008414.2 | 695 | 71 | ||
XM_015152929.2 | XP_015008415.2 | 695 | 71 | |||
GenBank mRNA | JV636235.1 | - | 562 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
