Assay Details
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143204.2 | XP_014998690.2 | 1982 | 75 | ||
GenBank mRNA | JU325095.1 | - | 963 | 75 | ||
JV044780.1 | - | 1491 | 75 | |||
JV705717.1 | - | 632 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
