This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07222600_m1
See other CRK GE Assays ›
Gene Symbol:
CRK
Gene Name:
CRK proto-oncogene, adaptor protein
Gene Aliases:
-
Chromosome Location:
Chr.16: 1273275 - 1308632 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07222600_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002808109.3 XP_002808155.2 473 108
XM_015118183.2 XP_014973669.1 473 108
GenBank mRNA JU333007.1 - 275 108
JU333008.1 - 275 108
JU333009.1 - 275 108
JU333010.1 - 275 108
JU333011.1 - 275 108
JU333012.1 - 275 108
JU333013.1 - 275 108
JU333014.1 - 275 108
JU333015.1 - 275 108
JU333016.1 - 275 108
JU474363.1 - 373 108
JU474364.1 - 373 108
JU474365.1 - 373 108
JU474366.1 - 373 108
JU474367.1 - 373 108
JV046235.1 - 374 108
JV046236.1 - 374 108
JV606373.1 - 306 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0