This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07222471_m1
See other GJA4 GE Assays ›
Gene Symbol:
GJA4
Gene Name:
gap junction protein alpha 4
Gene Aliases:
CXNH1
Chromosome Location:
Chr.1: 189838571 - 189841328 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07222471_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001108533.4 XP_001108533.2 103 66
GenBank mRNA JV680412.1 - 44 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001108533.4 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

gap junction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0