This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07219923_mH
See other LOC698860 GE Assays ›
Gene Symbol:
LOC698860
Gene Name:
general transcription factor II-I repeat domain-containing protein 2B
Gene Aliases:
GTF2IRD2, GTF2IRD2B
Chromosome Location:
Chr.3: 47155580 - 47217499 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07219923_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC698860
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028845698.1 XP_028701531.1 2422 63
XM_028845699.1 XP_028701532.1 2262 63
XM_028845700.1 XP_028701533.1 1079 63
GenBank mRNA JU334459.1 - 698 63
JU475548.1 - 614 63
JV043323.1 - 784 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Macaca mulatta

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0