Assay Details
Gene Symbol: PRPF38B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146394.2 | XP_015001880.1 | 2456 | 111 | ||
XM_028830944.1 | XP_028686777.1 | 2509 | 111 | |||
XM_028830945.1 | XP_028686778.1 | 2466 | 111 | |||
XM_028830949.1 | XP_028686782.1 | 2519 | 111 | |||
XM_028830952.1 | XP_028686785.1 | 2456 | 111 | |||
XM_028830954.1 | XP_028686787.1 | 2466 | 111 | |||
XM_028830955.1 | XP_028686788.1 | 2509 | 111 | |||
XM_028830957.1 | XP_028686790.1 | 2456 | 111 | |||
XM_028830962.1 | XP_028686795.1 | 2519 | 111 | |||
XM_028830968.1 | XP_028686801.1 | 2456 | 111 | |||
XM_028830970.1 | XP_028686803.1 | 2456 | 111 | |||
XM_028830974.1 | XP_028686807.1 | 2466 | 111 | |||
XM_028830975.1 | XP_028686808.1 | 2509 | 111 | |||
XM_028830977.1 | XP_028686810.1 | 2519 | 111 | |||
GenBank mRNA | JU321931.1 | - | 780 | 111 | ||
JU471133.1 | - | 702 | 111 | |||
JU489040.1 | - | 712 | 111 | |||
JV043657.1 | - | 757 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028830945.1, XM_028830949.1, XM_028830952.1, XM_028830954.1, XM_028830955.1, XM_028830957.1, XM_028830962.1, XM_028830968.1, XM_028830970.1, XM_028830974.1, XM_028830975.1, XM_028830977.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
