This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04959652_g1
See other COX7A2L GE Assays ›
Gene Symbol:
COX7A2L LOC106992283
Gene Name:
cytochrome c oxidase subunit VIIa polypeptide 2 like
cytochrome c oxidase subunit 7A-related protein, mitochondrial
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 50967740 - 50968859 on Build Mmul_10
Chromosome Location:
Chr.13: 66434462 - 66469979 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh04959652_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX7A2L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301704.1 NP_001288633.1 916 154
GenBank mRNA CN805580.1 - 195 154
JV044473.1 - 897 154
JV668573.1 - 893 154
Gene Symbol: LOC106992283
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015112234.2 XP_014967720.1 1143 154
XM_028831728.1 XP_028687561.1 1020 154
XM_028831729.1 XP_028687562.1 848 154

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta

Panther Classification:

Molecular Function -

oxidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0