This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04959564_m1
See other APOBEC3H GE Assays ›
Gene Symbol:
APOBEC3H
Gene Name:
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases:
-
Chromosome Location:
Chr.10: 12471710 - 12484284 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04959564_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOBEC3H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001345935.1 NP_001332864.1 214 98
XM_015150343.2 XP_015005829.2 270 98
XM_028827468.1 XP_028683301.1 255 98
XM_028827469.1 XP_028683302.1 342 98
GenBank mRNA DQ507277.1 - 151 98
JU481501.1 - 214 98
JU481502.1 - 214 98
JV637136.1 - 259 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Macaca mulatta

Panther Classification:

Molecular Function -

RNA processing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0