This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04958459_s1
See other CCR5 GE Assays ›
Gene Symbol:
CCR5
Gene Name:
C-C motif chemokine receptor 5
Gene Aliases:
EGK_11711
Chromosome Location:
Chr.2: 102388544 - 102394326 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04958459_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCR5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042773.3 NP_001036238.2 1224 69
NM_001309402.1 NP_001296331.1 1029 69
XM_015131082.2 XP_014986568.1 1248 69
GenBank mRNA DQ499963.1 - 1003 69
DQ499964.1 - 1003 69
DQ499965.1 - 1003 69
DQ499966.1 - 1003 69
DQ499967.1 - 1003 69
DQ499968.1 - 1003 69
DQ902484.1 - 1224 69
DQ902485.1 - 1224 69
DQ902486.1 - 1224 69
DQ902487.1 - 1224 69
DQ902488.1 - 1224 69
DQ902489.1 - 1224 69
DQ902490.1 - 1224 69
DQ902491.1 - 1224 69
DQ902492.1 - 1224 69
DQ902493.1 - 1224 69
DQ902494.1 - 1224 69
DQ902495.1 - 1224 69
DQ902496.1 - 1224 69
DQ902497.1 - 1224 69
DQ902498.1 - 1224 69
DQ902499.1 - 1224 69
DQ902500.1 - 1224 69
DQ902501.1 - 1224 69
DQ902502.1 - 1224 69
DQ902503.1 - 1224 69
DQ902504.1 - 1224 69
DQ902505.1 - 1224 69
DQ902506.1 - 1224 69
DQ902507.1 - 1224 69
DQ902508.1 - 1224 69
DQ902509.1 - 1224 69
DQ902510.1 - 1224 69
DQ902511.1 - 1224 69
DQ902512.1 - 1224 69
DQ902513.1 - 1224 69
DQ902514.1 - 1223 69
DQ902515.1 - 1224 69
DQ902516.1 - 1224 69
DQ902517.1 - 1224 69
DQ902518.1 - 1224 69
DQ902519.1 - 1224 69
DQ902520.1 - 1224 69
DQ902521.1 - 1224 69
DQ902522.1 - 1224 69
DQ902523.1 - 1224 69
DQ902524.1 - 1224 69
DQ902525.1 - 1224 69
DQ902526.1 - 1224 69
DQ902527.1 - 1224 69
DQ902528.1 - 1224 69
DQ902529.1 - 1224 69
DQ902530.1 - 1224 69
DQ902531.1 - 1224 69
DQ902532.1 - 1224 69
DQ902533.1 - 1224 69
DQ902534.1 - 1224 69
DQ902535.1 - 1224 69
DQ902536.1 - 1224 69
DQ902537.1 - 1224 69
DQ902538.1 - 69
DQ902539.1 - 1224 69
DQ902540.1 - 1224 69
DQ902541.1 - 1224 69
DQ902542.1 - 1224 69
DQ902543.1 - 1224 69
U73739.1 - 992 69
U96762.1 - 992 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0