Assay Details
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113249.2 | XP_014968735.1 | 267 | 74 | ||
GenBank mRNA | JU325478.1 | - | 29 | 74 | ||
JV541702.1 | - | 32 | 74 | |||
JV648716.1 | - | 70 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
