This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04383465_g1
See other GPR182 GE Assays ›
Gene Symbol:
GPR182
Gene Name:
G protein-coupled receptor 182
Gene Aliases:
ADMR
Chromosome Location:
Chr.11: 56557451 - 56561423 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04383465_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR182
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001115515.4 XP_001115515.3 196 117
XM_015152071.2 XP_015007557.2 196 117
XM_015152072.2 XP_015007558.2 196 117
XM_015152073.2 XP_015007559.2 196 117
XM_028829631.1 XP_028685464.1 196 117

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001115515.4, XM_015152071.2, XM_015152072.2, XM_015152073.2, XM_028829631.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0