This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04382637_m1
See other CRCP GE Assays ›
Gene Symbol:
CRCP
Gene Name:
CGRP receptor component
Gene Aliases:
-
Chromosome Location:
Chr.3: 53725261 - 53764667 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04382637_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRCP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015133381.2 XP_014988867.1 464 55
XM_015133382.2 XP_014988868.1 277 55
XM_028845718.1 XP_028701551.1 490 55
XM_028845720.1 XP_028701553.1 1138 55
GenBank mRNA JU334022.1 - 412 55
JU334023.1 - 412 55
JU334024.1 - 412 55
JU334025.1 - 412 55
JU475201.1 - 319 55
JV043255.1 - 412 55
JV578599.1 - 424 55
JV578600.1 - 424 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0