This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04372740_g1
See other ADGRA1 GE Assays ›
Gene Symbol:
ADGRA1
Gene Name:
adhesion G protein-coupled receptor A1
Gene Aliases:
GPR123
Chromosome Location:
Chr.9: 133551024 - 133593275 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04372740_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028827057.1 XP_028682890.1 632 92
GenBank mRNA JV566943.1 - 908 92
JV600802.1 - 921 92
JV667282.1 - 784 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0