This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04371395_m1
See other ZNF548 GE Assays ›
Gene Symbol:
ZNF548 ZNF547
Gene Name:
zinc finger protein 548
zinc finger protein 547
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 57006622 - 57017550 on Build Mmul_10
Chromosome Location:
Chr.19: 56982217 - 56998871 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh04371395_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF548
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002801418.3 XP_002801464.2 256 57
GenBank mRNA JU472961.1 - 87 57
JV052618.1 - 286 57
Gene Symbol: ZNF547
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA JV712622.1 - 177 57

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_002801418.3 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0