This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04366271_m1
See other MRO GE Assays ›
Gene Symbol:
MRO
Gene Name:
maestro
Gene Aliases:
-
Chromosome Location:
Chr.18: 31576777 - 31621775 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04366271_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015121976.2 XP_014977462.2 1120 89
XM_015121977.2 XP_014977463.2 1118 89
XM_015121978.2 XP_014977464.2 913 89
XM_015121980.2 XP_014977466.2 653 89
XM_015121985.2 XP_014977471.2 748 89
XM_015121986.2 XP_014977472.2 1004 89
XM_015121987.2 XP_014977473.2 2687 89
XM_028837777.1 XP_028693610.1 1051 89
XM_028837778.1 XP_028693611.1 1357 89
XM_028837779.1 XP_028693612.1 1007 89
XM_028837780.1 XP_028693613.1 886 89
XM_028837781.1 XP_028693614.1 905 89
GenBank mRNA JV045280.1 - 626 89
JV663026.1 - 626 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0