This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04344357_m1
See other PPP1R3C GE Assays ›
Gene Symbol:
PPP1R3C
Gene Name:
protein phosphatase 1 regulatory subunit 3C
Gene Aliases:
-
Chromosome Location:
Chr.9: 91816833 - 91821497 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04344357_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R3C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257737.1 NP_001244666.1 133 62
GenBank mRNA JU322787.1 - 133 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001257737.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0