This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04343820_g1
See other TRA2A GE Assays ›
Gene Symbol:
TRA2A
Gene Name:
transformer 2 alpha homolog
Gene Aliases:
-
Chromosome Location:
Chr.3: 91576679 - 91605794 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04343820_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRA2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258113.1 NP_001245042.1 1010 70
XM_015133842.2 XP_014989328.1 1643 70
XM_015133843.2 XP_014989329.1 1700 70
XM_015133844.2 XP_014989330.1 9808 70
XM_015133845.2 XP_014989331.1 1363 70
XM_015133846.1 XP_014989332.1 1352 70
XM_015133847.1 XP_014989333.1 1142 70
XM_028844741.1 XP_028700574.1 1249 70
XM_028844742.1 XP_028700575.1 863 70
GenBank mRNA JU475328.1 - 1010 70
JV046878.1 - 1017 70
JV046879.1 - 1011 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0