This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04256668_s1
See other PGAM1 GE Assays ›
Gene Symbol:
PGAM1
Gene Name:
phosphoglycerate mutase 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 97874020 - 97881905 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04256668_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195655.1 NP_001182584.1 47 75
GenBank mRNA CO648017.1 - 50 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001195655.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

mutase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0