This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04253248_g1
See other CYRIA GE Assays ›
Gene Symbol:
CYRIA
Gene Name:
CYFIP related Rac1 interactor A
Gene Aliases:
EGK_05069, FAM49A
Chromosome Location:
Chr.13: 92262706 - 92372290 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04253248_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYRIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193919.3 NP_001180848.1 779 66
XM_015111701.2 XP_014967187.1 905 66
XM_028831450.1 XP_028687283.1 906 66
XM_028831451.1 XP_028687284.1 1005 66
XM_028831452.1 XP_028687285.1 405 66
GenBank mRNA JU320018.1 - 862 66
JU320019.1 - 862 66
JU471251.1 - 800 66
JV452269.1 - 825 66
JV559193.1 - 839 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0