Assay Details
Gene Symbol: IER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002802002.3 | XP_002802048.3 | 421 | 57 | ||
GenBank mRNA | JU475870.1 | - | 215 | 57 | ||
JV618660.1 | - | 214 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002802002.3 transcripts.
Set Membership: |
![]() ![]() |
