This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh03043379_gH
See other ACTB GE Assays ›
Gene Symbol:
ACTB ACTG1 LOC705671
Gene Name:
actin beta
actin gamma 1
actin, cytoplasmic 2-like
Gene Aliases:
-
-
beta-actin
Chromosome Location:
Chr.3: 38643025 - 38646254 on Build Mmul_10
Chromosome Location:
Chr.16: 77852631 - 77855567 on Build Mmul_10
Chromosome Location:
Chr.2: 133462315 - 133463984 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
-
Assay ID Rh03043379_gH
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACTB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033084.1 NP_001028256.1 830 122
GenBank mRNA AY369786.1 - 49 122
Gene Symbol: ACTG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA JV667467.1 - 820 122
JV694889.1 - 820 122
Gene Symbol: LOC705671
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028844381.1 XP_028700214.1 754 122
GenBank mRNA AY497558.1 - 641 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0