This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02913019_m1
See other RHOG GE Assays ›
Gene Symbol:
RHOG
Gene Name:
ras homolog family member G
Gene Aliases:
-
Chromosome Location:
Chr.14: 64615716 - 64630149 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02913019_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RHOG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001113619.4 XP_001113619.1 642 103
GenBank mRNA JV047031.1 - 89 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001113619.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

small GTPase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0