This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02912365_m1
See other GPR34 GE Assays ›
Gene Symbol:
GPR34
Gene Name:
G protein-coupled receptor 34
Gene Aliases:
-
Chromosome Location:
Chr.X: 41556145 - 41565125 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02912365_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266567.1 NP_001253496.1 87 141
XM_015127182.2 XP_014982668.1 122 141
GenBank mRNA JU321287.1 - 87 141

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001266567.1, XM_015127182.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6889b944d9-flx6z:80/100.66.128.142:80.
git-commit: df865406a845caddbd23a531a4b79e504db49c75
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header