This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02893751_m1
See other CABP1 GE Assays ›
Gene Symbol:
CABP1
Gene Name:
calcium binding protein 1
Gene Aliases:
EGK_04269
Chromosome Location:
Chr.11: 120442123 - 120469371 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02893751_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CABP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015152969.2 XP_015008455.1 1083 74
XM_028829983.1 XP_028685816.1 947 74
XM_028829984.1 XP_028685817.1 672 74
XM_028829985.1 XP_028685818.1 672 74
XM_028829986.1 XP_028685819.1 346 74
GenBank mRNA JU325973.1 - 619 74
JV564977.1 - 473 74
JV598921.1 - 788 74
JV701817.1 - 787 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0