This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02891747_m1
See other DACT2 GE Assays ›
Gene Symbol:
DACT2
Gene Name:
dishevelled binding antagonist of beta catenin 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 2303452 - 2314319 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02891747_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DACT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001083736.4 XP_001083736.2 652 130
XM_015137719.2 XP_014993205.2 1741 130
XM_015137720.2 XP_014993206.2 1929 130
GenBank mRNA JV635990.1 - 476 130
JV708431.1 - 409 130

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015137719.2, XM_015137720.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0