Assay Details
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150089.2 | XP_015005575.2 | 6195 | 96 | ||
XM_015150090.2 | XP_015005576.2 | 6110 | 96 | |||
XM_015150091.2 | XP_015005577.2 | 6371 | 96 | |||
XM_015150092.2 | XP_015005578.2 | 6123 | 96 | |||
GenBank mRNA | JU333776.1 | - | 6074 | 96 | ||
JU475020.1 | - | 6184 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
