Assay Details
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172360.1 | NP_001165831.1 | 409 | 121 | ||
XM_015138044.2 | XP_014993530.2 | 926 | 121 | |||
XM_015138046.2 | XP_014993532.1 | 419 | 121 | |||
XM_028848137.1 | XP_028703970.1 | 992 | 121 | |||
XM_028848138.1 | XP_028703971.1 | 286 | 121 | |||
XM_028848139.1 | XP_028703972.1 | 638 | 121 | |||
XM_028848140.1 | XP_028703973.1 | 294 | 121 | |||
XM_028848141.1 | XP_028703974.1 | 568 | 121 | |||
XM_028848142.1 | XP_028703975.1 | 926 | 121 | |||
XM_028848144.1 | XP_028703977.1 | 472 | 121 | |||
XM_028848145.1 | XP_028703978.1 | 419 | 121 | |||
XM_028848146.1 | XP_028703979.1 | 415 | 121 | |||
XM_028848148.1 | XP_028703981.1 | 419 | 121 | |||
XM_028848149.1 | XP_028703982.1 | 419 | 121 | |||
XM_028848150.1 | XP_028703983.1 | 419 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028848140.1, XM_028848141.1, XM_028848142.1, XM_028848145.1, XM_028848146.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
