This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02880974_m1
See other ASPSCR1 GE Assays ›
Gene Symbol:
ASPSCR1
Gene Name:
ASPSCR1 tether for SLC2A4, UBX domain containing
Gene Aliases:
-
Chromosome Location:
Chr.16: 78282988 - 78326891 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02880974_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASPSCR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028837007.1 XP_028692840.1 1255 89
XM_028837008.1 XP_028692841.1 1240 89
XM_028837009.1 XP_028692842.1 1206 89
XM_028837010.1 XP_028692843.1 1255 89
XM_028837011.1 XP_028692844.1 1129 89
XM_028837012.1 XP_028692845.1 1255 89
XM_028837013.1 XP_028692846.1 1255 89
XM_028837014.1 XP_028692847.1 1129 89
GenBank mRNA JV628430.1 - 1206 89
JV667477.1 - 1161 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0