This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02876454_m1
See other LOC718510 GE Assays ›
Gene Symbol:
LOC718510
Gene Name:
calcium-binding protein 5
Gene Aliases:
CABP5, EGK_10826, PLA2G4C
Chromosome Location:
Chr.19: 48006074 - 48083664 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02876454_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC718510
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015124368.2 XP_014979854.1 488 99
XM_028838571.1 XP_028694404.1 2796 99
XM_028838572.1 XP_028694405.1 2796 99
XM_028838573.1 XP_028694406.1 2740 99
GenBank mRNA JV513295.1 - 33 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

phospholipase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0