This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02873916_m1
See other DET1 GE Assays ›
Gene Symbol:
DET1
Gene Name:
DET1 partner of COP1 E3 ubiquitin ligase
Gene Aliases:
EGK_17805
Chromosome Location:
Chr.7: 67287988 - 67312528 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02873916_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DET1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261519.1 NP_001248448.1 1191 93
XM_015142940.2 XP_014998426.1 1256 93
XM_028850235.1 XP_028706068.1 1242 93
XM_028850236.1 XP_028706069.1 1297 93
XM_028850237.1 XP_028706070.1 1297 93
XM_028850238.1 XP_028706071.1 1135 93
XM_028850239.1 XP_028706072.1 1112 93
GenBank mRNA JU320193.1 - 1149 93
JV043786.1 - 1191 93
JV635706.1 - 1198 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0