This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02871009_m1
See other LOC711104 GE Assays ›
Gene Symbol:
LOC711104
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
LUC7L2
Chromosome Location:
Chr.3: 165781692 - 165848890 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02871009_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC711104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001108444.4 XP_001108444.1 854 131
XM_015135024.2 XP_014990510.1 527 131
XM_015135025.2 XP_014990511.1 859 131
XM_015135026.2 XP_014990512.1 3209 131
XM_015135028.2 XP_014990514.1 2729 131
XM_015135034.2 XP_014990520.1 788 131
XM_015135036.2 XP_014990522.1 4313 131
XM_015135040.2 XP_014990526.1 924 131
XM_028846365.1 XP_028702198.1 1543 131
GenBank mRNA JU319762.1 - 741 131
JU471676.1 - 502 131
JV044892.1 - 660 131

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0