This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02868808_m1
See other LARP4B GE Assays ›
Gene Symbol:
LARP4B
Gene Name:
La ribonucleoprotein 4B
Gene Aliases:
EGK_19403
Chromosome Location:
Chr.9: 1531045 - 1659063 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02868808_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LARP4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260689.1 NP_001247618.1 1741 148
XM_015146331.2 XP_015001817.2 5330 148
XM_015146332.2 XP_015001818.2 5330 148
XM_015146335.2 XP_015001821.2 1753 148
XM_015146336.2 XP_015001822.2 1752 148
XM_028853540.1 XP_028709373.1 2004 148
GenBank mRNA JU333592.1 - 1736 148
JU471563.1 - 1740 148
JV046538.1 - 1741 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0