This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02863368_m1
See other OGFR GE Assays ›
Gene Symbol:
OGFR
Gene Name:
opioid growth factor receptor
Gene Aliases:
-
Chromosome Location:
Chr.10: 97964030 - 97973903 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02863368_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGFR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001087181.4 XP_001087181.3 887 86
XM_015148631.2 XP_015004117.2 405 86
XM_015148632.2 XP_015004118.2 606 86
XM_028828686.1 XP_028684519.1 882 86
XM_028828687.1 XP_028684520.1 887 86
XM_028828688.1 XP_028684521.1 404 86
GenBank mRNA JV604297.1 - 355 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0