Assay Details
Gene Symbol: IFI44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143215.2 | XP_014998701.2 | 1234 | 136 | ||
XM_015143220.2 | XP_014998706.2 | 1225 | 136 | |||
GenBank mRNA | JU478906.1 | - | 1233 | 136 | ||
JV049817.1 | - | 1212 | 136 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
