This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02855723_m1
See other GRAP2 GE Assays ›
Gene Symbol:
GRAP2
Gene Name:
GRB2 related adaptor protein 2
Gene Aliases:
EGK_03067
Chromosome Location:
Chr.10: 11567337 - 11645561 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02855723_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRAP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261710.1 NP_001248639.1 91
XM_015150331.2 XP_015005817.1 1108 91
XM_015150334.2 XP_015005820.1 3053 91
XM_015150335.2 XP_015005821.1 986 91
XM_015150337.2 XP_015005823.1 1104 91
XM_015150338.2 XP_015005824.1 479 91
XM_028827274.1 XP_028683107.1 1790 91
XM_028827275.1 XP_028683108.1 1041 91
XM_028827276.1 XP_028683109.1 1340 91
XM_028827277.1 XP_028683110.1 1254 91
XM_028827278.1 XP_028683111.1 3012 91
GenBank mRNA JU471070.1 - 91

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0