Assay Details
Gene Symbol: SLC16A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116662.4 | XP_001116662.1 | 782 | 95 | ||
XM_015152152.2 | XP_015007638.1 | 1392 | 95 | |||
XM_015152153.2 | XP_015007639.1 | 1486 | 95 | |||
XM_015152154.2 | XP_015007640.1 | 603 | 95 | |||
XM_015152155.2 | XP_015007641.1 | 581 | 95 | |||
XM_015152156.2 | XP_015007642.1 | 1152 | 95 | |||
XM_028829683.1 | XP_028685516.1 | 922 | 95 | |||
XM_028829684.1 | XP_028685517.1 | 910 | 95 | |||
GenBank mRNA | JU496230.1 | - | 110 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
