This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02849329_m1
See other ZCCHC7 GE Assays ›
Gene Symbol:
ZCCHC7
Gene Name:
zinc finger CCHC-type containing 7
Gene Aliases:
-
Chromosome Location:
Chr.15: 46787370 - 47025654 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02849329_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZCCHC7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261183.1 NP_001248112.1 1128 124
XM_015117405.2 XP_014972891.1 1252 124
XM_015117408.2 XP_014972894.1 1610 124
XM_015117409.2 XP_014972895.1 1122 124
GenBank mRNA JU336674.1 - 1207 124
JU473195.1 - 1256 124
JV045749.1 - 1128 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0