Assay Details
Gene Symbol: SH3GL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838596.1 | XP_028694429.1 | 550 | 145 | ||
XM_028838597.1 | XP_028694430.1 | 674 | 145 | |||
XM_028838598.1 | XP_028694431.1 | 1411 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
