Assay Details
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028852259.1 | XP_028708092.1 | 507 | 112 | ||
XM_028852261.1 | XP_028708094.1 | 609 | 112 | |||
XM_028852262.1 | XP_028708095.1 | 391 | 112 | |||
XM_028852266.1 | XP_028708099.1 | 507 | 112 | |||
GenBank mRNA | JU322537.1 | - | 488 | 112 | ||
JU322538.1 | - | 488 | 112 | |||
JU470522.1 | - | 372 | 112 | |||
JU470525.1 | - | 372 | 112 | |||
JU470526.1 | - | 372 | 112 | |||
JV045087.1 | - | 431 | 112 | |||
JV045088.1 | - | 431 | 112 | |||
JV582138.1 | - | 494 | 112 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
