This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02842020_m1
See other CD46 GE Assays ›
Gene Symbol:
CD46
Gene Name:
CD46 molecule
Gene Aliases:
-
Chromosome Location:
Chr.1: 65254867 - 65292645 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02842020_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD46
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195746.1 NP_001182675.1 679 141
XM_015116875.1 XP_014972361.1 850 141
XM_015116884.1 XP_014972370.1 850 141
XM_015116890.1 XP_014972376.1 719 141
XM_015116897.1 XP_014972383.1 850 141
XM_015116900.2 XP_014972386.1 850 141
XM_015116902.1 XP_014972388.1 850 141
XM_015116909.1 XP_014972395.1 619 141
XM_015116915.1 XP_014972401.1 850 141
GenBank mRNA FS722731.1 - 298 141
JU334542.1 - 841 141
JU334543.1 - 841 141
JU334544.1 - 841 141
JU475628.1 - 831 141
JU475629.1 - 831 141
JU475630.1 - 831 141
JV047079.1 - 832 141
JV047080.1 - 832 141
JV047081.1 - 832 141
U87922.1 - 679 141

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0