This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02839697_m1
See other RUNX1 GE Assays ›
Gene Symbol:
RUNX1
Gene Name:
RUNX family transcription factor 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 11515197 - 11791957 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02839697_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RUNX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001265962.1 NP_001252891.1 1167 68
XM_015132890.2 XP_014988376.1 1160 68
XM_015132891.2 XP_014988377.1 1018 68
XM_028844818.1 XP_028700651.1 1414 68
XM_028844819.1 XP_028700652.1 1171 68
GenBank mRNA JU471230.1 - 1167 68
JU471231.1 - 1167 68
JU471232.1 - 1167 68
JU477609.1 - 1128 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

Runt transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0