This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02839302_m1
See other AKT2 GE Assays ›
Gene Symbol:
AKT2
Gene Name:
AKT serine/threonine kinase 2
Gene Aliases:
EGK_10619
Chromosome Location:
Chr.19: 39436147 - 39489611 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02839302_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001265989.1 NP_001252918.1 640 70
XM_015123938.2 XP_014979424.1 703 70
XM_015123939.2 XP_014979425.1 851 70
XM_015123940.2 XP_014979426.1 667 70
XM_028838366.1 XP_028694199.1 2244 70
XM_028838367.1 XP_028694200.1 596 70
GenBank mRNA JU320025.1 - 690 70
JU471528.1 - 640 70
JV459925.1 - 459 70
JV644983.1 - 691 70

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0