This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02832248_g1
See other NIPA2 GE Assays ›
Gene Symbol:
NIPA2
Gene Name:
NIPA magnesium transporter 2
Gene Aliases:
EGK_17287
Chromosome Location:
Chr.7: 561517 - 596506 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02832248_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NIPA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194886.2 NP_001181815.1 888 105
XM_001106204.3 XP_001106204.1 1069 105
XM_015141678.2 XP_014997164.1 933 105
XM_028850397.1 XP_028706230.1 1320 105
GenBank mRNA JU321211.1 - 1029 105
JU471015.1 - 1029 105
JU471016.1 - 893 105
JV045757.1 - 1025 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0