Assay Details
Gene Symbol: ATP5F1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266412.1 | NP_001253341.1 | 232 | 116 | ||
GenBank mRNA | JV452960.1 | - | 249 | 116 | ||
JV565380.1 | - | 232 | 116 | |||
JV600174.1 | - | 232 | 116 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
