This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02830485_m1
See other COP1 GE Assays ›
Gene Symbol:
COP1
Gene Name:
COP1 E3 ubiquitin ligase
Gene Aliases:
RFWD2
Chromosome Location:
Chr.1: 33206875 - 33463769 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02830485_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266265.1 NP_001253194.1 1444 103
XM_015123338.2 XP_014978824.1 1777 103
XM_015123346.2 XP_014978832.1 1729 103
XM_015123351.2 XP_014978837.1 1717 103
XM_015123363.2 XP_014978849.1 1592 103
XM_028843468.1 XP_028699301.1 1843 103
XM_028843469.1 XP_028699302.1 1831 103
XM_028843470.1 XP_028699303.1 1783 103
GenBank mRNA JU327299.1 - 682 103
JU476785.1 - 1384 103
JU476786.1 - 1432 103
JU476787.1 - 1444 103
JV554568.1 - 788 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0