This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02826930_m1
See other EDNRB GE Assays ›
Gene Symbol:
EDNRB
Gene Name:
endothelin receptor type B
Gene Aliases:
-
Chromosome Location:
Chr.17: 59709777 - 59734504 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02826930_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EDNRB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001089047.4 XP_001089047.3 1614 125
XM_015121270.2 XP_014976756.1 2016 125
GenBank mRNA JU321243.1 - 1458 125
JV708730.1 - 1370 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0