This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02825840_m1
See other MRPL55 GE Assays ›
Gene Symbol:
MRPL55
Gene Name:
mitochondrial ribosomal protein L55
Gene Aliases:
-
Chromosome Location:
Chr.1: 85681462 - 85686200 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02825840_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001085425.4 XP_001085425.1 2597 102
XM_015114606.2 XP_014970092.1 483 102
XM_028826399.1 XP_028682232.1 430 102
XM_028826400.1 XP_028682233.1 968 102
XM_028826405.1 XP_028682238.1 971 102
GenBank mRNA JU323493.1 - 426 102
JU472806.1 - 429 102
JV045739.1 - 459 102
JV639105.1 - 429 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0