This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02822707_m1
See other CDKN1A GE Assays ›
Gene Symbol:
CDKN1A
Gene Name:
cyclin dependent kinase inhibitor 1A
Gene Aliases:
EGK_14841
Chromosome Location:
Chr.4: 132755001 - 132763844 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02822707_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKN1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194722.2 NP_001181651.1 662 66
XM_015136157.2 XP_014991643.1 675 66
XM_015136158.2 XP_014991644.1 653 66
XM_028846890.1 XP_028702723.1 792 66
XM_028846891.1 XP_028702724.1 1154 66
XM_028846892.1 XP_028702725.1 1680 66
GenBank mRNA JU478686.1 - 294 66
JV568672.1 - 570 66
JV602692.1 - 560 66
JV667323.1 - 517 66

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 3' Most Endogenous Controls Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

protein-binding activity modulator kinase inhibitor kinase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0